Novus Biologicals
Manufacturer Code:NBP15653620UL
Catalog # NBP15653620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to UBQLN1(ubiquilin 1) The peptide sequence was selected from the middle region of UBQLN1. Peptide sequence QFGGNPFASLVSNTSSGEGSQPSRTENRDPLPNPWAPQTSQSSSASSGTA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DA41FLJ90054 DSK2 hPLIC-1 PLIC1 PLIC-1UBQN Protein linking IAP with cytoskeleton 1 ubiquilin 1 ubiquilin-1 XDRP1; hPLIC-1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; Ubiquilin-1
Gene Aliases: DA41; DSK2; PLIC-1; PLIC1; UBQLN1; UBQN; XDRP1
UniProt ID: (Human) Q9UMX0
Entrez Gene ID: (Human) 29979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.