Proteintech
Manufacturer Code:221261AP150UL
Catalog # 50-173-3733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Ubiquilin 1 Fusion Protein Ag17530 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DA41 DSK2 hPLIC 1 PLIC 1 PLIC1 ubiquilin 1 UBQLN1 XDRP1; hPLIC-1; PLIC-1; Protein linking IAP with cytoskeleton 1; testicular tissue protein Li 219; Ubiquilin-1
Gene Aliases: DA41; DSK2; PLIC-1; PLIC1; UBQLN1; UBQN; XDRP1
UniProt ID: (Human) Q9UMX0, (Rat) Q9JJP9
Entrez Gene ID: (Human) 29979, (Rat) 114590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.