Novus Biologicals
Manufacturer Code:NBP300032UV
Catalog # NB064058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant UbcH8/Ube2L6 (O14933-1 Met1-Ser153). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 ubiquitin-conjugating enzyme L6; EC 6.3.2.19 retinoic acid induced gene B protein Retinoic acid-induced gene B protein RIG-B UbcH8 UBCH8MGC40331 Ubiquitin carrier protein L6 ubiquitin/ISG15-conjugating enzyme E2 L6 ubiquitin-conjugating enzyme E2L 6 Ubiquitin-protein ligase L6; retinoic acid induced gene B protein; Retinoic acid-induced gene B protein; RIG-B; UbcH8; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2L 6; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6; Ubiquitin/ISG15-conjugating enzyme E2 L6
Gene Aliases: RIG-B; UBCH8; UBE2L6
UniProt ID: (Human) O14933
Entrez Gene ID: (Human) 9246
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.