Invitrogen
Manufacturer Code:PA599733
Catalog # PIPA599733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Uba5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730525G14Rik AW240750 FLJ23251 FLJ23251UBA5 THIFP1 Uba5 UBA5 ubiquitin-activating enzyme E1 homolog UBE1DC1 ubiquitin like modifier activating enzyme 5 ubiquitin-activating enzyme 5 Ubiquitin-activating enzyme E1 domain-containing protein 1 ubiquitin-activating enzyme E1-domain containing 1 ubiquitin-like modifier activating enzyme 5 ubiquitin-like modifier-activating enzyme 5 UFM1-activating enzyme; ThiFP1; UBA5, ubiquitin-activating enzyme E1 homolog; Ubiquitin-activating enzyme 5; Ubiquitin-activating enzyme E1 domain-containing protein 1; ubiquitin-activating enzyme E1-domain containing 1; ubiquitin-like modifier activating enzyme 5; Ubiquitin-like modifier-activating enzyme 5; UFM1-activating enzyme
Gene Aliases: 5730525G14Rik; AW240750; THIFP1; UBA5; UBE1DC1
UniProt ID: (Human) Q9GZZ9, (Rat) Q5M7A4, (Mouse) Q8VE47
Entrez Gene ID: (Human) 79876, (Rat) 300968, (Mouse) 66663
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.