Invitrogen
Manufacturer Code:PA526082
Catalog # PIPA526082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1247-1276 amino acids from the C-terminal region of human UTY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H3]-trimethyl-L-lysine(27) demethylase UTY; Histone demethylase UTY; male-specific histocompatibility antigen H-YDB; ubiquitous TPR motif protein UTY; ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome; ubiquitously transcribed tetratricopeptide repeat gene, Y-linked; ubiquitously transcribed TPR gene on Y chromosome; ubiquitously transcribed TPR protein on the Y chromosome; ubiquitously transcribed Y chromosome tetratricopeptide repeat protein; Ubiquitously-transcribed TPR protein on the Y chromosome; Ubiquitously-transcribed Y chromosome tetratricopeptide repeat protein; UTY1 histone demethylase UTY tetratricopeptide repeat protein ubiquitous TPR motif protein UTY ubiquitously transcribed TPR gene on Y chromosome ubiquitously transcribed tetratricopeptide repeat gene Y chromosome ubiquitously-transcribed TPR protein on the Y
Gene Aliases: Hydb; KDM6AL; KDM6C; mKIAA4057; UTY; UTY1
UniProt ID: (Human) O14607
Entrez Gene ID: (Human) 7404, (Mouse) 22290
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.