Invitrogen
Manufacturer Code:PA5114160
Catalog # PIPA5114160
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal region of human UTP23 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700010I21Rik AI662478 BB238373 C8orf53 D530033C11Rik RGD1307179 rRNA-processing protein UTP23 homolog Utp23 UTP23 small subunit (SSU) processome component homolog UTP23 small subunit (SSU) processome component homolog (yeast) UTP23 small subunit processome component; rRNA-processing protein UTP23 homolog; UTP23, small subunit (SSU) processome component, homolog
Gene Aliases: C8orf53; UTP23
UniProt ID: (Human) Q9BRU9
Entrez Gene ID: (Human) 84294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.