Invitrogen
Manufacturer Code:PA590468
Catalog # PIPA590468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 70-260 of human UTP18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: U3 small nucleolar RNA-associated protein 18 homolog; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; UTP18, small subunit processome component; WD repeat domain 50; WD repeat-containing protein 50
Gene Aliases: 6230425C22Rik; 6430513M13; CDABP0061; CGI-48; UTP18; WDR50
UniProt ID: (Human) Q9Y5J1, (Mouse) Q5SSI6
Entrez Gene ID: (Human) 51096, (Mouse) 217109
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.