Novus Biologicals
Manufacturer Code:NBP297492AF532
Catalog # NB102267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human UTP18 fragment. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-48 U3 small nucleolar RNA-associated protein 18 homolog UTP18 small subunit (SSU) processome component homolog (yeast) WD repeat domain 50 WD repeat-containing protein 50 WDR50; U3 small nucleolar RNA-associated protein 18 homolog; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; WD repeat domain 50; WD repeat-containing protein 50
Gene Aliases: CDABP0061; CGI-48; UTP18; WDR50
UniProt ID: (Human) Q9Y5J1
Entrez Gene ID: (Human) 51096
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.