Novus Biologicals
Manufacturer Code:NB10077295
Catalog # NB10077295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human small subunit (SSU) processome component UTP18 (WD repeat domain 50) using the numbering given in entry NP_057085.2 (GeneID 51096). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-48 U3 small nucleolar RNA-associated protein 18 homolog UTP18 small subunit (SSU) processome component homolog (yeast) WD repeat domain 50 WD repeat-containing protein 50 WDR50; U3 small nucleolar RNA-associated protein 18 homolog; UTP18 small subunit (SSU) processome component homolog; UTP18 small subunit processome component; UTP18, small subunit (SSU) processome component, homolog; WD repeat domain 50; WD repeat-containing protein 50
Gene Aliases: CDABP0061; CGI-48; UTP18; WDR50
UniProt ID: (Human) Q9Y5J1
Entrez Gene ID: (Human) 51096
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.