Invitrogen
Manufacturer Code:PA5113210
Catalog # PIPA5113210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human U3 small nucleolar RNA-associated protein 15 homolog protein (234-518AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW544865 BOS_19659 NET21 RGD1310992 Src-associated protein SAW U3 small nucleolar RNA-associated protein 15 homolog UniProtKB:Q8TED0} UTP15 utp15 {ECO:0000250 UTP15 SSU processome component UTP15 small subunit processome component UTP15 U3 small nucleolar ribonucleoprotein homolog UTP15 U3 small nucleolar ribonucleoprotein homolog (S. cerevisiae) UTP15 U3 small nucleolar ribonucleoprotein homolog (yeast); Src-associated protein SAW; U3 small nucleolar RNA-associated protein 15 homolog; UTP15 SSU processome component; UTP15, U3 small nucleolar ribonucleoprotein, homolog
Gene Aliases: NET21; UTP15
UniProt ID: (Human) Q8TED0
Entrez Gene ID: (Human) 84135
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.