Proteintech
Manufacturer Code:174161AP150UL
Catalog # 50-173-1564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UTP15 Fusion Protein Ag11161 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET21 UTP15; Src-associated protein SAW; U3 small nucleolar RNA-associated protein 15 homolog; UTP15 SSU processome component; utp15 {ECO:0000250|UniProtKB:Q8TED0}; UTP15, U3 small nucleolar ribonucleoprotein, homolog
Gene Aliases: AW544865; NET21; RGD1310992; UTP15
UniProt ID: (Human) Q8TED0, (Rat) A2RRU3, (Mouse) Q8C7V3
Entrez Gene ID: (Human) 84135, (Rat) 310019, (Mouse) 105372
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.