Proteintech
Manufacturer Code:114741AP150UL
Catalog # 50-172-7191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | UTP14A Fusion Protein Ag2027 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY CO 16 KIAA0266 NY CO 16 SDCCAG16 UTP14A; Antigen NY-CO-16; Jsd-like X-linked protein; Juvenile spermatogonial depletion-like X-linked protein; Serologically defined colon cancer antigen 16; U3 small nucleolar RNA-associated protein 14 homolog A; UTP14, U3 small nucleolar ribonucleoprotein, homolog A; UTP14A small subunit (SSU) processome component
Gene Aliases: 2700066J21Rik; dJ537K23.3; JsdX; Kiaa0266; mKIAA0266; NY-CO-16; NYCO16; SDCCAG16; T25628; UTP14A
UniProt ID: (Human) Q9BVJ6, (Mouse) Q640M1
Entrez Gene ID: (Human) 10813, (Mouse) 72554, (Rat) 317579
Molecular Function: RNA binding protein nuclease nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.