Novus Biologicals
Manufacturer Code:NBP274823F
Catalog # NBP274823F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F3 |
Immunogen | Full length recombinant protein of human UTP11L (NP_057121) produced in E.coli. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CGI94 CGI-94 probable U3 small nucleolar RNA-associated protein 11 U3 snoRNA-associated protein 11 UTP11-like protein UTP11-like U3 small nucleolar ribonucleoprotein (yeast); Probable U3 small nucleolar RNA-associated protein 11; U3 snoRNA-associated protein 11; UTP11-like protein; UTP11-like, U3 small nucleolar ribonucleoprotein
Gene Aliases: CGI-94; CGI94; HDCMB12P; UTP11; UTP11L
UniProt ID: (Human) Q9Y3A2
Entrez Gene ID: (Human) 51118
Molecular Function:
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.