Invitrogen
Manufacturer Code:PA519122
Catalog # PIPA519122
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PSPPKSADASPAP) corresponding to the internal amino acids of UTF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Undifferentiated embryonic cell transcription factor 1; UTF1; UTF1 undifferentiated embryonic cell transcription factor 1 OTTHUMP00000020786
Gene Aliases: UTF1
UniProt ID: (Human) Q5T230
Entrez Gene ID: (Human) 8433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.