Novus Biologicals
Manufacturer Code:NBP29377420UL
Catalog # NB029411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 420-580 of human SLC14A2 (NP_009094.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ16167 HUT2UT-A2 hUT-A6 MGC119566 solute carrier family 14 (urea transporter) member 2 Solute carrier family 14 member 2 urea transporter 2 Urea transporter kidney urea transporter-2 UT2MGC119567 UTA UTR; solute carrier family 14 (urea transporter), member 2; Solute carrier family 14 member 2; Urea transporter 2; Urea transporter, kidney; urea transporter-2
Gene Aliases: hUT-A6; HUT2; SLC14A2; UT-A2; UT2; UTA; UTR
UniProt ID: (Human) Q15849
Entrez Gene ID: (Human) 8170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.