Invitrogen
Manufacturer Code:PA577786
Catalog # PIPA577786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced against a synthetic peptide mapped to the C-terminal tail (amino acids 911-929) of rat UT-A1 (antibody designation L194) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: plasma membrane urea transporter; SLC14A2 FLJ16167 hUT-A6 HUT2 kidney MGC119566 MGC119567 Slc14a2 Solute carrier family 14 (urea transporter) member 2 Solute carrier family 14 member 2 Urea transporter 2 Urea transporter Urea transporter kidney UT-A2 UT2 UT2&lowbarHUMAN UTA UTR UT1 UTA1; solute carrier family 14 (urea transporter), member 2, variant 4; solute carrier family 14 (urea transporter), member 2T; Solute carrier family 14 member 2; solute carrier family 14, member 2; testName; urea transport protein; Urea transporter 2; Urea transporter, kidney
Gene Aliases: Slc14a2; Slc14a2_v4; Slc14a2T; testSymbol; UrT1-C; UrT1-D; UT-A1; UT-A3; Ut2
UniProt ID: (Rat) Q62668, (Mouse) Q8R4T9
Entrez Gene ID: (Rat) 54302, (Mouse) 27411
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.