Invitrogen
Manufacturer Code:PA5116409
Catalog # PIPA5116409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 42-76 amino acids from the human region of human SLC14A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ16167 HUT2 hUT-A6 MGC119566 MGC119567 plasma membrane urea transporter Slc14a2 Slc14a2_v4 Slc14a2T solute carrier family 14 (urea transporter) member 2 solute carrier family 14 (urea transporter) member 2 variant 4 solute carrier family 14 (urea transporter) member 2T solute carrier family 14 member 2 solute carrier family 14 member 2 testName testSymbol urea transport protein urea transporter 2 Urea transporter kidney urea transporter-2 UrT1-C UrT1-D UT1 Ut2 UTA UTA1 UT-A1 UT-A2 UT-A3 UTR; solute carrier family 14 (urea transporter), member 2; Solute carrier family 14 member 2; Urea transporter 2; Urea transporter, kidney; urea transporter-2
Gene Aliases: hUT-A6; HUT2; SLC14A2; UT-A2; UT2; UTA; UTR
UniProt ID: (Human) Q15849
Entrez Gene ID: (Human) 8170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.