Novus Biologicals
Manufacturer Code:NBP275704
Catalog # NBP275704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG35-11 |
Immunogen | Recombinant protein within human USP9X aa 1-200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Deubiquitinating enzyme FAF-X; Deubiquitinating enzyme FAF-X DFFRXubiquitin specific protease 9 X-linked (fat facets-like Drosophila) Drosophila fat facets related X-linked EC 3.1.2.15 EC 3.4.19.12 FAF FAM Fat facets in mammals fat facets protein related X-linked Fat facets protein-related X-linked hFAM probable ubiquitin carboxyl-terminal hydrolase FAF-X ubiquitin specific peptidase 9 X-linked ubiquitin specific peptidase 9 X-linked (fat facets-like Drosophila) ubiquitin thioesterase FAF-X Ubiquitin thiolesterase FAF-X ubiquitin-specific processing protease FAF-X Ubiquitin-specific protease 9 X chromosome Ubiquitin-specific-processing protease FAF-X USP9 X chromosome (fat facets-like Drosophila); Drosophila fat facets related, X-linked; Fat facets in mammals; fat facets protein related, X-linked; Fat facets protein-related, X-linked; hFAM; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); Ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; Ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X
Gene Aliases: DFFRX; FAF; FAM; MRX99; MRXS99F; USP9; USP9X
UniProt ID: (Human) Q93008
Entrez Gene ID: (Human) 8239
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.