Invitrogen
Manufacturer Code:PA570428
Catalog # PIPA570428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human USP9Y |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme FAF-Y; Fat facets protein-related, Y-linked; probable ubiquitin carboxyl-terminal hydrolase FAF-X; Probable ubiquitin carboxyl-terminal hydrolase FAF-Y; ubiquitin specific peptidase 9, Y chromosome; ubiquitin specific peptidase 9, Y-linked; ubiquitin specific peptidase 9, Y-linked (fat facets-like, Drosophila); ubiquitin specific protease 9, Y chromosome; Ubiquitin thioesterase FAF-Y; ubiquitin thiolesterase FAF-Y; ubiquitin-specific processing protease FAF-Y; Ubiquitin-specific protease 9, Y chromosome; Ubiquitin-specific-processing protease FAF-Y; USP9Y DFFRY
Gene Aliases: DFFRY; Fafl2; SPGFY2; USP9Y
UniProt ID: (Human) O00507
Entrez Gene ID: (Bovine) 100271721, (Human) 8287, (Rat) 103694559, (Mouse) 107868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.