Invitrogen
Manufacturer Code:MA526367
Catalog # PIMA526367
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2246-2570 of USP9X produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme FAF-X; DFFRX FAF FAM Drosophila fat facets related X-linked deubiquitinating enzyme FAF-X fat facets in mammals fat facets protein related X-linked fat facets protein-related X-linked hFAM probable ubiquitin carboxyl-terminal hydrolase FAF-X ubiquitin specific protease 9 X chromosome (fat facets-like Drosophila) ubiquitin thioesterase FAF-X ubiquitin thiolesterase FAF-X ubiquitin-specific processing protease FAF-X ubiquitin-specific protease 9 X chromosome ubiquitin-specific-process; Drosophila fat facets related, X-linked; Fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; Fat facets protein-related, X-linked; fats facets protein related, X; hFAM; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-X; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); Ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X; Ubiquitin-specific protease 9, X chromosome; Ubiquitin-specific-processing protease FAF-X
Gene Aliases: 5730589N07Rik; AA407302; AA407699; AL022658; AL022749; DFFRX; FAF; FAF-X; Fafl; FAM; MRX99; MRXS99F; RGD1560056; USP9; USP9X
UniProt ID: (Human) Q93008, (Mouse) P70398
Entrez Gene ID: (Dog) 480885, (Rhesus monkey) 106992260, (Human) 8239, (Mouse) 22284, (Rat) 363445
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.