Proteintech
Manufacturer Code:277911AP150UL
Catalog # 50-173-5760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP8 Fusion Protein Ag27143 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 8; Deubiquitinating enzyme 8 hUBPy HumORF8 KIAA0055 Ubiquitin isopeptidase Y ubiquitin specific peptidase 8 Ubiquitin thiolesterase 8 UBPY USP8; hUBPy; mUBPy; putative deubiquitinating enzyme; Ubiquitin carboxyl-terminal hydrolase 8; Ubiquitin isopeptidase Y; Ubiquitin thioesterase 8; ubiquitin thiolesterase 8; Ubiquitin-specific-processing protease 8
Gene Aliases: AI574262; AW557536; HumORF8; KIAA0055; mKIAA0055; SPG59; UBPY; USP8
UniProt ID: (Human) P40818, (Mouse) Q80U87
Entrez Gene ID: (Human) 9101, (Mouse) 84092
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.