Invitrogen
Manufacturer Code:PA517179
Catalog # PIPA517179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues surrounding Asp438 of human HAUSP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 7; Herpes virus-associated ubiquitin-specific protease; Herpes virus-associated ubiquitin-specific protease deubiquitinating enzyme 7 herpesvirus-associated ubiquitin-specific protease ubiquitin carboxyl-terminal hydrolase 7 ubiquitin specific protease 7 (herpes virus-associated) ubiquitin thioesterase 7 ubiquitin-specific-processing protease 7; herpes-virus-associated ubiquitin-specific protease; Herpesvirus-associated ubiquitin-specific protease; mHAUSP; rHAUSP; Ubiquitin carboxyl-terminal hydrolase 7; ubiquitin specific protease 7; ubiquitin specific protease 7 (herpes virus-associated); Ubiquitin thioesterase 7; ubiquitin thiolesterase 7; Ubiquitin-specific-processing protease 7
Gene Aliases: 2210010O09Rik; AA409944; AA617399; AU019296; AW548146; C80752; HAUSP; TEF1; USP7
UniProt ID: (Human) Q93009, (Rat) Q4VSI4, (Mouse) Q6A4J8
Entrez Gene ID: (Human) 7874, (Rat) 360471, (Mouse) 252870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.