Novus Biologicals
Manufacturer Code:NBP274813AF405
Catalog # N274813A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G6 |
Immunogen | Recombinant protein fragment corresponding to amino acids 358-700 of human USP54 (NP_689799) produced in E.coli. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: bA137L10.3 bA137L10.4 C10orf29 USP54 ubiquitin specific peptidase 54; Inactive ubiquitin carboxyl-terminal hydrolase 54; Inactive ubiquitin-specific peptidase 54; ubiquitin specific protease 54; ubiquitin specific proteinase 54
Gene Aliases: bA137L10.3; bA137L10.4; C10orf29; USP54
UniProt ID: (Human) A1L4Q4
Entrez Gene ID: (Human) 159195
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.