Novus Biologicals
Manufacturer Code:NBP2860480.1ML
Catalog # NB033977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human USP54 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA137L10.3 bA137L10.4 C10orf29 USP54 ubiquitin specific peptidase 54; Inactive ubiquitin carboxyl-terminal hydrolase 54; Inactive ubiquitin-specific peptidase 54; ubiquitin specific protease 54; ubiquitin specific proteinase 54
Gene Aliases: bA137L10.3; bA137L10.4; C10orf29; USP54
UniProt ID: (Human) A1L4Q4
Entrez Gene ID: (Human) 159195
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.