Abnova
Manufacturer Code:PAB28659
Catalog # 89-130-171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human USP51. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant USP51.
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 51; Ubiquitin carboxyl-terminal hydrolase 51; Ubiquitin thioesterase 51; ubiquitin thiolesterase 51; Ubiquitin-specific-processing protease 51
Gene Aliases: USP51
UniProt ID: (Human) Q70EK9
Entrez Gene ID: (Human) 158880
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.