Invitrogen
Manufacturer Code:PA5100214
Catalog # PIPA5100214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human UBP5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA407472 Deubiquitinating enzyme 5 isopeptidase T ISOT ISOT-1 testicular tissue protein Li 218 ubiquitin carboxy terminal hydrolase T ubiquitin carboxyl-terminal hydrolase 5 ubiquitin isopeptidase T ubiquitin specific peptidase 5 ubiquitin specific peptidase 5 (isopeptidase T) ubiquitin specific protease 5 ubiquitin specific protease 5 (isopeptidase T) ubiquitin thioesterase 5 ubiquitin thiolesterase 5 ubiquitin-specific protease-5 (ubiquitin isopeptidase T) ubiquitin-specific-processing protease 5 Ucht Usp5; Deubiquitinating enzyme 5; Isopeptidase T; testicular tissue protein Li 218; ubiquitin carboxy terminal hydrolase T; Ubiquitin carboxyl-terminal hydrolase 5; ubiquitin isopeptidase T; ubiquitin specific peptidase 5 (isopeptidase T); ubiquitin specific protease 5; ubiquitin specific protease 5 (isopeptidase T); Ubiquitin thioesterase 5; ubiquitin thiolesterase 5; ubiquitin-specific protease-5 (ubiquitin isopeptidase T); Ubiquitin-specific-processing protease 5
Gene Aliases: AA407472; ISOT; ISOT-1; Ucht; USP5
UniProt ID: (Human) P45974, (Mouse) P56399
Entrez Gene ID: (Human) 8078, (Rat) 297593, (Mouse) 22225
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.