Invitrogen
Manufacturer Code:PA530661
Catalog # PIPA530661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1186 and 1375 of Human USP47 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 47; Trf (TATA binding protein-related factor)-proximal homolog; TRFP Trf (TATA binding protein-related factor)-proximal homolog deubiquitinating enzyme 47 ubiquitin carboxyl-terminal hydrolase 47 ubiquitin specific protease 47 ubiquitin thioesterase 47 ubiquitin thiolesterase 47 ubiquitin-specific-processing protease 47; Ubiquitin carboxyl-terminal hydrolase 47; ubiquitin specific protease 47; Ubiquitin thioesterase 47; ubiquitin thiolesterase 47; Ubiquitin-specific-processing protease 47
Gene Aliases: 4930502N04Rik; A630020C16Rik; TRFP; USP47
UniProt ID: (Human) E9PM46, (Mouse) Q8BY87
Entrez Gene ID: (Human) 55031, (Mouse) 74996
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.