Novus Biologicals
Manufacturer Code:NBP220836
Catalog # NBP220836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human USP47. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 47; Deubiquitinating enzyme 47 DKFZp686C13257 EC 3.4.19.12 FLJ20727 Trf (TATA binding protein-related factor)-proximal homolog Trf (TATA binding protein-related factor)-proximal homolog (Drosophila) TRFP ubiquitin carboxyl-terminal hydrolase 47 ubiquitin specific peptidase 47 ubiquitin specific protease 47 ubiquitin thioesterase 47 Ubiquitin thiolesterase 47 Ubiquitin-specific-processing protease 47; Trf (TATA binding protein-related factor)-proximal homolog; Ubiquitin carboxyl-terminal hydrolase 47; Ubiquitin thioesterase 47; ubiquitin thiolesterase 47; Ubiquitin-specific-processing protease 47
Gene Aliases: TRFP; USP47
UniProt ID: (Human) E9PM46
Entrez Gene ID: (Human) 55031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.