Abnova
Manufacturer Code:H00055031A01
Catalog # 89-107-003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 203 a.a. &bsim 301 a.a) partial recombinant protein with GST tag.,USP47 (NP_060414 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant USP47.
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 47; DKFZp686C13257; FLJ20727; Trf (TATA binding protein-related factor)-proximal homolog; TRFP; Ubiquitin carboxyl-terminal hydrolase 47; Ubiquitin thioesterase 47; ubiquitin thiolesterase 47; Ubiquitin-specific-processing protease 47
Gene Aliases: TRFP; USP47
UniProt ID: (Human) E9PM46
Entrez Gene ID: (Human) 55031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.