OriGene
Manufacturer Code:CF801911
Catalog # 50-166-5776
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 420-712 of human USP44 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 44; Ubiquitin carboxyl-terminal hydrolase 44; ubiquitin specific peptidase 44; ubiquitin specific protease 44; Ubiquitin thioesterase 44; ubiquitin thiolesterase 44; Ubiquitin-specific-processing protease 44
Gene Aliases: USP44
UniProt ID: (Human) Q9H0E7
Entrez Gene ID: (Human) 84101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.