Novus Biologicals
Manufacturer Code:NBP29318720UL
Catalog # NB028262
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-270 of human USP44 (NP_115523.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 44; Deubiquitinating enzyme 44 DKFZp434D0127 EC 3.1.2.15 EC 3.4.19.12 FLJ14528 ubiquitin carboxyl-terminal hydrolase 44 ubiquitin specific peptidase 44 ubiquitin specific protease 44 ubiquitin thioesterase 44 Ubiquitin thiolesterase 44 Ubiquitin-specific-processing protease 44; Ubiquitin carboxyl-terminal hydrolase 44; ubiquitin specific protease 44; Ubiquitin thioesterase 44; ubiquitin thiolesterase 44; Ubiquitin-specific-processing protease 44
Gene Aliases: USP44
UniProt ID: (Human) Q9H0E7
Entrez Gene ID: (Human) 84101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.