Novus Biologicals
Manufacturer Code:NB1002869
Catalog # NB1002869
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 925 and the C-terminus (residue 963) of human Ubiquitin Specific Protease 4 using the numbering given in entry AAB72237.1 (GeneID 7375). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 4; Deubiquitinating enzyme 4 EC 3.1.2.15 EC 3.4.19.12 MGC149848 MGC149849 ubiquitin carboxyl-terminal esterase 4 ubiquitin carboxyl-terminal hydrolase 4 ubiquitin specific peptidase 4 (proto-oncogene) ubiquitin specific protease 4 (proto-oncogene) ubiquitin thioesterase 4 Ubiquitin thiolesterase 4 ubiquitin-specific processing protease 4 Ubiquitin-specific-processing protease 4 Ubiquitous nuclear protein homolog UNPH UNPUnph; ubiquitin carboxyl-terminal esterase 4; Ubiquitin carboxyl-terminal hydrolase 4; ubiquitin specific peptidase 4 (proto-oncogene); ubiquitin specific protease 4 (proto-oncogene); Ubiquitin thioesterase 4; ubiquitin thiolesterase 4; ubiquitin-specific processing protease 4; Ubiquitin-specific-processing protease 4; Ubiquitous nuclear protein homolog
Gene Aliases: UNP; UNPH; USP4
UniProt ID: (Human) Q13107
Entrez Gene ID: (Human) 7375
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.