Invitrogen
Manufacturer Code:MA524903
Catalog # PIMA524903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D11 |
Immunogen | Full length human recombit protein of USP38 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 38; HP43.8KD; KIAA1891; Ubiquitin carboxyl-terminal hydrolase 38; ubiquitin specific protease 38; Ubiquitin thioesterase 38; ubiquitin thiolesterase 38; Ubiquitin-specific-processing protease 38
Gene Aliases: HP43.8KD; KIAA1891; USP38
UniProt ID: (Human) Q8NB14
Entrez Gene ID: (Human) 84640, (Dog) 476071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.