Proteintech
Manufacturer Code:177671AP150UL
Catalog # 50-173-1778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP38 Fusion Protein Ag12172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 38; Deubiquitinating enzyme 38 HP43.8KD KIAA1891 Ubiquitin thiolesterase 38 USP38; HP43.8KD; Ubiquitin carboxyl-terminal hydrolase 38; ubiquitin specific protease 38; Ubiquitin thioesterase 38; ubiquitin thiolesterase 38; Ubiquitin-specific-processing protease 38
Gene Aliases: 4631402N15Rik; 4833420O05Rik; AA536967; AU044701; AW544820; HP43.8KD; KIAA1891; mKIAA1891; USP38
UniProt ID: (Human) Q8NB14, (Mouse) Q8BW70
Entrez Gene ID: (Human) 84640, (Mouse) 74841, (Rat) 307764
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.