OriGene
Manufacturer Code:TA810971
Catalog # 50-168-3739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H4 |
Immunogen | Full length human recombit protein of human USP25 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 25; Ubiquitin carboxyl-terminal hydrolase 25; Ubiquitin thioesterase 25; ubiquitin thiolesterase 25; ubiquitin-specific processing protease 25; Ubiquitin-specific-processing protease 25; USP on chromosome 21; USP21
Gene Aliases: USP21; USP25
UniProt ID: (Human) Q9UHP3
Entrez Gene ID: (Human) 29761
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.