Novus Biologicals
Manufacturer Code:NBP180631
Catalog # NBP180631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QPLVGIETLPPDLRDFVEEDNQRFEKELEEWDAQLAQKALQEKLLASQKLRESETSVTTAQAAGDPEYLEQPSRSDFSKHLKEETIQIITKASHEHEDKSPETVLQSIMMTPNMQGIIMAIGKSRSVYDRCGPEAGFFK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 25; Deubiquitinating enzyme 25 EC 3.1.2.15 EC 3.4.19.12 ubiquitin carboxyl-terminal hydrolase 25 ubiquitin specific peptidase 25 ubiquitin specific protease USP2510USP21ubiquitin specific protease 25 ubiquitin thioesterase 25 Ubiquitin thiolesterase 25 ubiquitin-specific processing protease 25 Ubiquitin-specific-processing protease 25 USP on chromosome 21; Ubiquitin carboxyl-terminal hydrolase 25; Ubiquitin thioesterase 25; ubiquitin thiolesterase 25; ubiquitin-specific processing protease 25; Ubiquitin-specific-processing protease 25; USP on chromosome 21
Gene Aliases: USP21; USP25
UniProt ID: (Human) Q9UHP3
Entrez Gene ID: (Human) 29761
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.