Proteintech
Manufacturer Code:131261AP150UL
Catalog # 50-172-8509
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP24 Fusion Protein Ag3779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 24; Deubiquitinating enzyme 24 KIAA1057 Ubiquitin thiolesterase 24 USP24; Ubiquitin carboxyl-terminal hydrolase 24; ubiquitin specific protease 24; Ubiquitin thioesterase 24; ubiquitin thiolesterase 24; ubiquitin-specific processing protease 24; Ubiquitin-specific-processing protease 24
Gene Aliases: 2700066K03Rik; 2810030C21Rik; AI414051; B130021E18; C79851; KIAA1057; USP24
UniProt ID: (Human) Q9UPU5, (Mouse) B1AY13
Entrez Gene ID: (Human) 23358, (Mouse) 329908
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.