Invitrogen
Manufacturer Code:PA531634
Catalog # PIPA531634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 189 and 524 of Human USP22 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 22; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific protease 22; Ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; Ubiquitin-specific-processing protease 22; USP3L deubiquitinating enzyme 22 ubiquitin carboxyl-terminal hydrolase 22 ubiquitin specific protease 22 ubiquitin thioesterase 22 ubiquitin thiolesterase 22 ubiquitin-specific processing protease 22 ubiquitin-specific-processing protease 22
Gene Aliases: KIAA1063; USP22; USP3L
UniProt ID: (Human) Q9UPT9
Entrez Gene ID: (Human) 23326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.