Novus Biologicals
Manufacturer Code:NBP149644
Catalog # NBP149644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to an internal region of the human USP22 protein (within residues 100-205). [Swiss-Prot Q9H3R0] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 22; Deubiquitinating enzyme 22 EC 3.1.2.15 EC 3.4.19.12 KIAA1063ubiquitin carboxyl-terminal hydrolase 22 KIAA1064 ubiquitin specific peptidase 22 ubiquitin specific peptidase 3-like ubiquitin specific protease 22 ubiquitin thioesterase 22 Ubiquitin thiolesterase 22 ubiquitin-specific processing protease 22 Ubiquitin-specific-processing protease 22 USP3L; Ubiquitin carboxyl-terminal hydrolase 22; ubiquitin specific protease 22; Ubiquitin thioesterase 22; ubiquitin thiolesterase 22; ubiquitin-specific processing protease 22; Ubiquitin-specific-processing protease 22
Gene Aliases: KIAA1063; USP22; USP3L
UniProt ID: (Human) Q9UPT9
Entrez Gene ID: (Human) 23326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.