Proteintech
Manufacturer Code:174911AP150UL
Catalog # 50-173-1602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP20 Fusion Protein Ag11590 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 20; Deubiquitinating enzyme 20 hVDU2 KIAA1003 LSFR3A Ubiquitin thiolesterase 20 USP20 VDU2; hVDU2; pVHL-interacting deubiquitinating enzyme 2; Ubiquitin carboxyl-terminal hydrolase 20; ubiquitin specific protease 20; Ubiquitin thioesterase 20; ubiquitin thiolesterase 20; Ubiquitin-specific-processing protease 20; VHL-interacting deubiquitinating enzyme 2
Gene Aliases: 1700055M05Rik; AI467231; hVDU2; KIAA1003; LSFR3A; USP20; VDU2
UniProt ID: (Human) Q9Y2K6, (Mouse) Q8C6M1
Entrez Gene ID: (Human) 10868, (Mouse) 74270, (Rat) 311856
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.