Proteintech
Manufacturer Code:103921AP150UL
Catalog # 50-172-6270
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP2 Fusion Protein Ag0591 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 41 kDa ubiquitin-specific protease; Deubiquitinating enzyme 2; Deubiquitinating enzyme 2 ubiquitin specific peptidase 2 Ubiquitin thiolesterase 2 UBP41 USP2 USP9; Ubiquitin carboxyl-terminal hydrolase 2; ubiquitin specific protease 12; ubiquitin specific protease 2; ubiquitin specific protease 9; Ubiquitin thioesterase 2; ubiquitin thiolesterase 2; Ubiquitin-specific-processing protease 2; Ubiquitin-specific-processing protease testis; UBP-t
Gene Aliases: B930035K21Rik; MNCb-0190; UBP-t; UBP41; Ubp69; USP2; USP9
UniProt ID: (Human) O75604, (Mouse) O88623, (Rat) Q5U349
Entrez Gene ID: (Human) 9099, (Mouse) 53376, (Rat) 115771
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.