Novus Biologicals
Manufacturer Code:NBP29352920UL
Catalog # NB028934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human USP19 (NP_001186089.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 19; Deubiquitinating enzyme 19 EC 3.1.2.15 EC 3.4.19.12 KIAA0891 ubiquitin carboxyl-terminal hydrolase 19 ubiquitin specific peptidase 19 ubiquitin thioesterase 19 Ubiquitin thiolesterase 19 Ubiquitin-specific-processing protease 19 Zinc finger MYND domain-containing protein 9 ZMYND9ubiquitin specific protease 19; Ubiquitin carboxyl-terminal hydrolase 19; ubiquitin specific protease 19; Ubiquitin thioesterase 19; ubiquitin thiolesterase 19; Ubiquitin-specific-processing protease 19; Zinc finger MYND domain-containing protein 9
Gene Aliases: KIAA0891; USP19; ZMYND9
UniProt ID: (Human) O94966
Entrez Gene ID: (Human) 10869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.