Proteintech
Manufacturer Code:257681AP150UL
Catalog # 50-173-5065
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP19 Fusion Protein Ag22646 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 19; Deubiquitinating enzyme 19 KIAA0891 Ubiquitin thiolesterase 19 USP19 ZMYND9; Ubiquitin carboxyl-terminal hydrolase 19; ubiquitin specific protease 19; Ubiquitin thioesterase 19; ubiquitin thiolesterase 19; Ubiquitin-specific-processing protease 19; Zinc finger MYND domain-containing protein 9
Gene Aliases: KIAA0891; USP19; ZMYND9
UniProt ID: (Human) O94966
Entrez Gene ID: (Human) 10869
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.