Invitrogen
Manufacturer Code:PA542843
Catalog # PIPA542843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human USP18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 43 kDa ISG15-specific protease; hUBP43; ISG15-specific-processing protease; ubiquitin specific protease 15; ubiquitin specific protease 18; Ubl carboxyl-terminal hydrolase 18; Ubl thioesterase 18; ubl thiolesterase 18
Gene Aliases: 1110058H21Rik; AW047653; bubp43; ISG43; Ubp15; UBP43; USP18
UniProt ID: (Human) Q9UMW8, (Mouse) Q9WTV6
Entrez Gene ID: (Bovine) 515202, (Horse) 100054488, (Rabbit) 100355501, (Dog) 486763, (Human) 11274, (Guinea pig) 100729301, (Mouse) 24110, (Rat) 312688
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.