Invitrogen
Manufacturer Code:PA544961
Catalog # PIPA544961
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human USP17L2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 17-like protein 2; deubiquitinating enzyme 3; Deubiquitinating protein 3; DUB-3; Ubiquitin carboxyl-terminal hydrolase 17; Ubiquitin carboxyl-terminal hydrolase 17-like protein 2; ubiquitin specific peptidase 17-like 2; Ubiquitin thioesterase 17-like protein 2; ubiquitin thiolesterase 17-like protein 2; Ubiquitin-specific-processing protease 17-like protein 2; USP17
Gene Aliases: DUB-3; DUB3; USP17; USP17H; USP17I; USP17J; USP17K; USP17L; USP17L2; USP17M
UniProt ID: (Human) Q6R6M4
Entrez Gene ID: (Human) 377630
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.