Proteintech
Manufacturer Code:140551AP150UL
Catalog # 50-172-9177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | USP16 Fusion Protein Ag4981 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 16; Deubiquitinating enzyme 16 Ubiquitin thiolesterase 16 UBP M USP16; deubiquitinating enzyme 16 {ECO:0000255|HAMAP-Rule:MF_03062}; Ubiquitin carboxyl-terminal hydrolase 16; ubiquitin carboxyl-terminal hydrolase 16 {ECO:0000255|HAMAP-Rule:MF_03062}; ubiquitin specific protease 16; Ubiquitin thioesterase 16; ubiquitin thioesterase 16 {ECO:0000255|HAMAP-Rule:MF_03062}; ubiquitin thiolesterase 16; Ubiquitin-processing protease UBP-M; ubiquitin-specific processing protease 16; Ubiquitin-specific-processing protease 16; ubiquitin-specific-processing protease 16 {ECO:0000255|HAMAP-Rule:MF_03062}; USP16
Gene Aliases: 1200004E02Rik; 2810483I07Rik; 6330514E22Rik; MSTP039; UBP-M; UBPM; USP16
UniProt ID: (Human) Q9Y5T5, (Rat) Q2KJ09, (Mouse) Q99LG0
Entrez Gene ID: (Human) 10600, (Rat) 288306, (Mouse) 74112
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.