Invitrogen
Manufacturer Code:PA521589
Catalog # PIPA521589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 920 and 981 of USP15 (Uniprot IDNo.Q9Y4E8) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 15; granule cell antiserum positive 18; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; UNPH-2 UNPH4 deubiquitinating enzyme 15 ubiquitin carboxyl-terminal hydrolase 15 ubiquitin thioesterase 15 ubiquitin thiolesterase 15 ubiquitin-specific-processing protease 15; Unph4
Gene Aliases: 4921514G19Rik; AI327321; E430033I05Rik; Gcap18; KIAA0529; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y4E8, (Mouse) Q8R5H1
Entrez Gene ID: (Human) 9958, (Mouse) 14479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.