Invitrogen
Manufacturer Code:PA5110706
Catalog # PIPA5110706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human USP15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921514G19Rik AI327321 BOS_5267 Deubiquitinating enzyme 15 E430033I05Rik Gcap18 Kiaa0529 MGC131982 MGC149838 MGC74854 Ubiquitin carboxyl-terminal hydrolase 15 Ubiquitin carboxyl-terminal hydrolase of 109 kDa Ubiquitin thioesterase 15 Ubiquitin-specific-processing protease 15 ubp109 Unph-2 Unph4 Usp15; Deubiquitinating enzyme 15; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; Unph4
Gene Aliases: KIAA0529; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y4E8
Entrez Gene ID: (Human) 9958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.