Invitrogen
Manufacturer Code:PA5101942
Catalog # PIPA5101942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human USP15 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921514G19Rik AI327321 Deubiquitinating enzyme 15 deubiquitinating enzyme Ubp109 E430033I05Rik Gcap18 granule cell antiserum positive 18 KIAA0529 MGC131982 MGC149838 MGC74854 Ubiquitin carboxyl-terminal hydrolase 15 ubiquitin carboxyl-terminal hydrolase of 109 kDa ubiquitin specific peptidase 15 ubiquitin specific protease 15 ubiquitin thioesterase 15 ubiquitin thiolesterase 15 Ubiquitin-specific-processing protease 15 Ubp109 UNPH-2 Unph4 USP15; Deubiquitinating enzyme 15; deubiquitinating enzyme Ubp109; granule cell antiserum positive 18; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin carboxyl-terminal hydrolase of 109 kDa; ubiquitin specific protease 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; Unph4
Gene Aliases: 4921514G19Rik; AI327321; E430033I05Rik; Gcap18; KIAA0529; Ubp109; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y4E8, (Mouse) Q8R5H1, (Rat) Q9R085
Entrez Gene ID: (Human) 9958, (Mouse) 14479, (Rat) 171329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.