Novus Biologicals
Manufacturer Code:NB1001255
Catalog # NB1001255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence NDIENENCMHTN corresponding to C-Terminus according to NP_006304.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating enzyme 15; Deubiquitinating enzyme 15 EC 3.1.2.15 EC 3.4.19.12 KIAA0529ubiquitin carboxyl-terminal hydrolase 15 MGC131982 MGC149838 MGC74854 ubiquitin specific peptidase 15 ubiquitin specific protease 15 ubiquitin thioesterase 15 Ubiquitin thiolesterase 15 Ubiquitin-specific-processing protease 15 unph-2 UNPH4; Ubiquitin carboxyl-terminal hydrolase 15; Ubiquitin thioesterase 15; ubiquitin thiolesterase 15; Ubiquitin-specific-processing protease 15; Unph-2; Unph4
Gene Aliases: KIAA0529; UNPH-2; UNPH4; USP15
UniProt ID: (Human) Q9Y4E8
Entrez Gene ID: (Human) 9958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.